Anti-BBS5 Rabbit Polyclonal Antibody

Supplier: Proteintech
14569-1-AP-150UL
10083-528EA 537.7 USD
10083-528
Anti-BBS5 Rabbit Polyclonal Antibody
Antibodies
BBS5 encodes a protein that has been directly linked to Bardet-Biedl syndrome. Bardet-Biedl syndrome (BBS) is an autosomal recessive condition characterised by rod-cone dystrophy, postaxial polydactyly, central obesity, mental retardation, hypogonadism, and renal dysfunction. Other associated clinical findings in BBS patients include diabetes, hypertension and congenital heart defects. BBS expression varies both within and between families and diagnosis is often difficult. Experimentation in non-human eukaryotes suggests that BBS5 is expressed in ciliated cells and that it is required for the formation of cilia. Alternate transcriptional splice variants have been observed but have not been fully characterized.

Western Blot: Mouse Testis Tissue, 1:200-1:1000; IF: HTERT-RPE1 Cells, 1:10-1:100; IHC: Human Testis Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: BBS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR