Anti-JIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14568-1-AP-150UL
10089-076EA 402 USD
10089-076
Anti-JIP1 Rabbit Polyclonal Antibody
Antibodies
IHC: Mouse Brain Tissue, 1:20-1:200; Western Blot:fetal Human Brain Tissue, 1:200-1:1000; IF:PC-12 Cells, 1:10-1:100

Type: Primary
Antigen: JIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR