Anti-CLIC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14545-1-AP-150UL
10084-860EA 402 USD
10084-860
Anti-CLIC1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human small intestine Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; Western Blot: HL-60 Cells, 1:200-1:2000

Type: Primary
Antigen: CLIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR