Anti-LMP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14544-1-AP-150UL
10089-738EA 402 USD
10089-738
Anti-LMP2 Rabbit Polyclonal Antibody
Antibodies
PSMB9, also named as LMP2, PSMB6i and RING12, belongs to the peptidase T1B family. The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. Replacement of PSMB6 by PSMB9 increases the capacity of the immunoproteasome to cleave model peptides after hydrophobic and basic residues. High expression of PSMB9 associated with the poor prognosis of patients with NPC.

Western Blot: Raji Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: LMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR