Anti-CEP250 Rabbit Polyclonal Antibody

Supplier: Proteintech
14498-1-AP-150UL
10084-646EA 537.7 USD
10084-646
Anti-CEP250 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: CEP250
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR