Anti-Gastrokine 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14494-1-AP-150UL
10087-406EA 402 USD
10087-406
Anti-Gastrokine 1 Rabbit Polyclonal Antibody
Antibodies
Gastrokine 1 (GKN1), a stomach-specific protein also known as 18 kDa antrum mucosa protein (AMP-18) or foveolin, belongs to the gastrokine family of gastric mucus cell-secreted proteins. The human GKN1 gene has been localized in a region of chromosome 2p13 of about 6 kb and contains 6 exons. GKN1 is expressed only in normal human stomach, but is absent from gastric adenocarcinomas, gastro-esophageal adenocarcinoma cell lines, and other normal and tumor gastro-intestinal tissues. GKN1 may play an important role in normal gastric function and may be a gastric tumour suppressor.

Western Blot: Human Stomach Tissue, 1:1000-1:10000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: Gastrokine 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR