Anti-VAPB Rabbit Polyclonal Antibody

Supplier: Proteintech
14477-1-AP-150UL
10096-922EA 537.7 USD
10096-922
Anti-VAPB Rabbit Polyclonal Antibody
Antibodies
Vesicle-associated membrane protein-associated protein B (VAPB) is an integral membrane protein localized to the endoplasmic reticulum (ER) membrane. VAPB has been implicated in various cellular processes, including ER stress, the unfolded protein response (UPR) and calcium homeostasis regulation. The mutations in the gene of VAPB cause amyotrophic lateral sclerosis 8 (ALS8) and some other related forms of motor neuron disease including late onset spinal muscular atrophy.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: VAPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR