Anti-LCOR Rabbit Polyclonal Antibody

Supplier: Proteintech
14476-1-AP-150UL
10089-558EA 402 USD
10089-558
Anti-LCOR Rabbit Polyclonal Antibody
Antibodies
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box . It functions through recruitment of C-terminal binding proteins (CtBPs) and histone deacetylases (HDACs) to its N-terminal and central domains, respectively . It is a scaffold for both histone deacetylases (HDACs) and C-terminal binding protein (CtBP) corepressors and functions by both HDAC-dependent and -independent mechanisms in a receptor-specific manner.

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human endometrial cancer, 1:20-1:200

Type: Primary
Antigen: LCOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR