Anti-BBS9 Rabbit Polyclonal Antibody

Supplier: Proteintech
14460-1-AP-150UL
10083-532EA 402 USD
10083-532
Anti-BBS9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human Liver Tissue, 1:20-1:200; IF: HTERT-RPE1 Cells, 1:10-1:100

Type: Primary
Antigen: BBS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR