Anti-CD276 Rabbit Polyclonal Antibody

Supplier: Proteintech
14453-1-AP-150UL
10084-308EA 402 USD
10084-308
Anti-CD276 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human medulloblastoma Tissue, 1:20-1:200; FC: RAW 264.7 Cells, N/A

Type: Primary
Antigen: CD276
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR