Anti-TSHR Rabbit Polyclonal Antibody

Supplier: Proteintech
14450-1-AP-150UL
10096-410EA 402 USD
10096-410
Anti-TSHR Rabbit Polyclonal Antibody
Antibodies
Thyroid stimulating hormone (also known as thyrotropin or TSH) is a glycoprotein hormone produced by thyrotrophs in the anterior pituitary gland. The TSH receptor (TSHR), mainly expressed on the thyroid epithelial cells, is a G protein-coupled receptor (GPCR) of the seven-transmembrane domain family and plays a central role in controlling thyroid cell metabolism. The gene of TSHR maps to chromosome 14q31.1, and encodes a 764-amino acid protein with a predicted unmodified molecular weight of 86.8 kDa. The apparent molecular weight of glycosylated form is 95-120 kDa. Studies reveal that TSHR is composed of two subunits, A and B, which are produced by cleavage of single-chain TSHR on the cell surface and subsequently connected by disulfide bonds. This rabbit polyclonal antibody raised against N-terminal region (1-253 aa) of human TSHR can detect subunit A with an experimentally determined molecular weight of 62-70 kDa under reducing condition.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: TSHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR