Anti-Orail-L2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14443-1-AP-150UL
10091-600EA 402 USD
10091-600
Anti-Orail-L2 Rabbit Polyclonal Antibody
Antibodies
Store-operated calcium channels (SOCs) play essential functions in various physiological processes. Orai1 (also named CRACM1) is a critical molecular component of SOCs. Orai1 protein is a 35-45 kDa plasma membrane protein with 4 transmembrane domains. Human patients with a severe combinaed immunodeficiency (SCID) syndrome possess point mutations in Orai1 gene. This antibody may be used to block the ion currents mediated by Orai1.

Western Blot: Human spleen Tissue, 1:200-1:1000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: Orail-L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR