Anti-APOA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14427-1-AP-150UL
10083-004EA 402 USD
10083-004
Anti-APOA1 Rabbit Polyclonal Antibody
Antibodies
ApoA1 is a major protein component of high density lipoproteins (HDL) which is associated with reversed cholesterol transport, lipid/cholesterol binding, lecithin/cholesterol acyltransferase (LCAT) activation and specific receptors binding. It is synthesized in the liver and small intestine. Defects of ApoA1 cause low HDL level and systemic non-neuropathic amyloidosis. Serum concentration of ApoA1 is inversely related to the risk of developing atherosclerosis. This antibody was generated against the C-terminal region of human ApoA1.

Western Blot: Human Blood Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR