Anti-KDM5C Rabbit Polyclonal Antibody

Supplier: Proteintech
14426-1-AP-150UL
10089-160EA 524.48 USD
10089-160
Anti-KDM5C Rabbit Polyclonal Antibody
Antibodies
KDM5C belongs to the ARID protein family that harbors conserved DNA-binding motifs and acts as a histone H3 lysine 4 demethylase, suggesting a potential role in epigenetic regulation during development, cell growth and differentiation KDM5C protein is homologous to three other proteins of the human JARID1 family (JARID1A, JARID1B and JARID1C). This family shares several evolutionarily conserved domains, including the catalytic JmjC domain, JmjN domain, Arid/Bright domain, C5HC2 zinc-finger domain and two plant homeodomains (PHD), a motif that binds methyl-lysine residues .

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: KDM5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR