Anti-CHSY1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14420-1-AP-150UL
10084-774EA 402 USD
10084-774
Anti-CHSY1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse spleen Tissue, 1:500-1:5000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: CHSY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR