Anti-GALE Rabbit Polyclonal Antibody

Supplier: Proteintech
14414-1-AP-150UL
10087-328EA 402 USD
10087-328
Anti-GALE Rabbit Polyclonal Antibody
Antibodies
GALE(Galactowaldenase) is also named as UDP-glucose 4-epimerase, UDP-galactose 4-epimerase, and belongs to the sugar epimerase family. UDP-galactose 4-epimerase catalyzes the interconversion of UDP-glucose and UDP-galactose during normal galactose metabolism. In humans, deficiencies in this enzyme lead to the complex disorder referred to as epimerase-deficiency galactosemia.It can exsit as a homodimer.

Western Blot:A375 Cells, 1:200-1:1000; IHC: Human malignant melanoma Tissue, 1:20-1:200

Type: Primary
Antigen: GALE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR