Anti-PIBF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14413-1-AP-150UL
10092-238EA 402 USD
10092-238
Anti-PIBF1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100; IP: Hea Cells, 1:200-1:2000

Type: Primary
Antigen: PIBF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR