Anti-HE4 Rabbit Polyclonal Antibody

Supplier: Proteintech
14406-1-AP-150UL
10088-136EA 402 USD
10088-136
Anti-HE4 Rabbit Polyclonal Antibody
Antibodies
WFDC2, also named as HE4, is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. HE4 is expressed in a number of normal tissues, and it is also highly expressed in a number of tumors cells lines, such ovarian, colon, breast, lung and renal cells lines. In some study, HE4 was referred to as a biomarker for ovarian carcinoma.

IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: HE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR