Anti-PSMA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14377-1-AP-150UL
10092-890EA 524.48 USD
10092-890
Anti-PSMA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: PSMA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR