Anti-RBM3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14363-1-AP-150UL
10093-412EA 402 USD
10093-412
Anti-RBM3 Rabbit Polyclonal Antibody
Antibodies
RBM3, also named as RNPL, is a cold-inducible mRNA binding protein that enhances global protein synthesis at both physiological and mild hypothermic temperatures. It reduces the relative abundance of microRNAs, when overexpressed. RBM3 enhances phosphoryaltion of translation initiation factors and active polysome formation. It is up-regulated in human tumors. RBM3 is a potential proto-oncogenic proteins upon overexpression. This antibody can bind the close sequences genes.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: RBM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR