Anti-ERG Rabbit Polyclonal Antibody

Supplier: Proteintech
14356-1-AP-150UL
10086-618EA 402 USD
10086-618
Anti-ERG Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF-7 Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:10-1:100

Type: Primary
Antigen: ERG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR