Anti-CRCP Rabbit Polyclonal Antibody

Supplier: Proteintech
14348-1-AP-150UL
10085-164EA 402 USD
10085-164
Anti-CRCP Rabbit Polyclonal Antibody
Antibodies
Western Blot: Raji Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: CRCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR