Anti-KIF4A Rabbit Polyclonal Antibody

Supplier: Proteintech
14344-1-AP-150UL
10089-238EA 402 USD
10089-238
Anti-KIF4A Rabbit Polyclonal Antibody
Antibodies
IHC: Human lung cancer, 1:20-1:200; IF: HepG2, 1:20-1:200

Type: Primary
Antigen: KIF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR