Anti-MAVS Rabbit Polyclonal Antibody

Supplier: Proteintech
14341-1-AP-150UL
10090-000EA 402 USD
10090-000
Anti-MAVS Rabbit Polyclonal Antibody
Antibodies
Mitochondrial antiviral-signaling protein (MAVS) is also known as virus-induced-signaling adapter (VISA) or interferon beta promoter stimulator protein 1 (IPS-1), it is widely involved and required for innate immune defense against viruses. MAVS, present in T cells, monocytes, epithelial cells and hepatocytes, contains CARD and transmembrane domains which are essential for antiviral functions. MAVS is able to interact with various cellular proteins including DDX58/RIG-I, IFIH1/MDA5, TRAF2, TRAF6, TMEM173/MITA, IFIT3 and etc. It can undergoe phosphorylation on multiple sites and ubiquitination, which may together cause the molecular weight migrate to about 70 kDa despite the predicated 57 kDa.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100; IP: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: MAVS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR