Anti-KIF3C Rabbit Polyclonal Antibody

Supplier: Proteintech
14333-1-AP-150UL
10089-236EA 402 USD
10089-236
Anti-KIF3C Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: KIF3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR