Anti-COG6 Rabbit Polyclonal Antibody

Supplier: Proteintech
14277-1-AP-150UL
10084-972EA 402 USD
10084-972
Anti-COG6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IF: MCF-7 Cells, 1:10-1:100; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: COG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR