Anti-SRXN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14273-1-AP-150UL
10095-194EA 524.48 USD
10095-194
Anti-SRXN1 Rabbit Polyclonal Antibody
Antibodies
IP:A549 Cells, 1:200-1:1000; IHC: Human lung cancer Tissue, 1:20-1:200; Western Blot:A549 Cells, 1:200-1:1000

Type: Primary
Antigen: SRXN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR