Anti-ITPKC Rabbit Polyclonal Antibody

Supplier: Proteintech
14267-1-AP-150UL
10089-050EA 402 USD
10089-050
Anti-ITPKC Rabbit Polyclonal Antibody
Antibodies
ITPKC(Inositol-trisphosphate 3-kinase C) is also named as IP3KC and belongs to the inositol phosphokinase (IPK) family. It can phosphorylate inositol 2,4,5-triphosphate to inositol 2,4,5,6-tetraphosphate and is relatively weakly activated by the calcium-calmodulin complex. ITPKC also acts as a negative regulator of T-cell activation through the Ca(2+)/NFAT signaling pathway, and the C allele may contribute to immune hyperreactivity in Kawasaki disease.

Western Blot: Human Heart Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: ITPKC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR