Anti-PNPLA6 Rabbit Polyclonal Antibody

Supplier: Proteintech
14261-1-AP-150UL
10092-524EA 462.47 USD
10092-524
Anti-PNPLA6 Rabbit Polyclonal Antibody
Antibodies
PNPLA6(patatin-like phospholipase domain-containing protein 6) is also named as NTE(neuropathy target esterase) and belongs to the NTE family. It plays an important role in motor neuron development and implicated overexpression of BMP signaling. Defects in PNPLA6 are the cause of spastic paraplegia autosomal recessive type 39 (SPG39). This protein has a glycosylaion site that it can be glycosylated. It has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IP:N/A, N/A

Type: Primary
Antigen: PNPLA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR