Anti-UBL4A Rabbit Polyclonal Antibody

Supplier: Proteintech
14253-1-AP-150UL
10096-674EA 402 USD
10096-674
Anti-UBL4A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: UBL4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR