Anti-ATX Rabbit Polyclonal Antibody

Supplier: Proteintech
14243-1-AP-150UL
10083-418EA 524.48 USD
10083-418
Anti-ATX Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ATX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR