Anti-MPI Rabbit Polyclonal Antibody

Supplier: Proteintech
14234-1-AP-150UL
10090-370EA 459.7 USD
10090-370
Anti-MPI Rabbit Polyclonal Antibody
Antibodies
MPI(mannose-6-phosphate isomerase) is also named as PMI1 and belongs to the mannose-6-phosphate isomerase type 1 family. It is a zinc metalloenzyme that catalyzes the reversible isomerization of mannose-6-phosphate and fructose-6-phosphate. MPI plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. This proterin has 2 isoforms produced by alternative splicing.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR