Anti-HSD11B2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14192-1-AP-150UL
10088-474EA 402 USD
10088-474
Anti-HSD11B2 Rabbit Polyclonal Antibody
Antibodies
The HSD11B2 gene encodes the type II isoform of 11-beta-hydroxysteroid dehydrogenase, a microsomal enzyme complex responsible for the interconversion of biologically active cortisol and inactive cortisone. This protein catalyzes the cortisol-to-cortisone reaction. This protein expresses approximately 44 kDa in placenta, trophoblast, and distal colon, but in kidney tissue, there are two bands of approximately 44 and 48 kDa can be consistently observed and no signal is seen in decidua, adrenal, or liver.

Western Blot: Human Kidney Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: HSD11B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR