Anti-CHD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
14173-1-AP-150UL
10084-694EA 402 USD
10084-694
Anti-CHD4 Rabbit Polyclonal Antibody
Antibodies
Chromodomain helicase DNA-binding protein 4 (CHD4) is an ATP dependent chromatin remodeler and a major subunit of the repressive NuRD complex . This complex plays key roles in transcriptional regulation and reorganization and maintenance of chromatin structure, as it targets pericentromeric heterochromatin and has recently been implicated in DNA damage repair, being rapidly recruited to DNA double strand breaks . CHD4 is a novel BRIT1 binding partner that regulates the HR repair process, which is a critical pathway of repairing DNA double strand breaks and plays an essential role in maintaining genomic integrity .

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200; IF:A549, 1:20-1:200

Type: Primary
Antigen: CHD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR