Anti-FKBP5 Rabbit Polyclonal Antibody

Supplier: Proteintech
14155-1-AP-150UL
10087-036EA 524.34 USD
10087-036
Anti-FKBP5 Rabbit Polyclonal Antibody
Antibodies
FKBP5(Peptidyl-prolyl cis-trans isomerase FKBP5 ) interacts with functionally mature heterooligomeric progesterone receptor complexes along with HSP90 and TEBP. Unlike the predominant T-cell expression of the mouse protein, FKBP5 is expressed as a 51-kD protein in a variety of tissues but not in brain, colon, or lung.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: FKBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR