Anti-RPL17 Rabbit Polyclonal Antibody

Supplier: Proteintech
14121-1-AP-150UL
10093-526EA 524.48 USD
10093-526
Anti-RPL17 Rabbit Polyclonal Antibody
Antibodies
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL17 gene encodes a ribosomal protein that is a component of the 60S subunit belonging to the L22P family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as rpL23 because the encoded protein shares amino acid identity with ribosomal protein L23 from Halobacterium marismortui; however, its official symbol is RPL17. The specific function of this protein is not well known to date.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IP: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: RPL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR