Anti-MARCH8 Rabbit Polyclonal Antibody

Supplier: Proteintech
14119-1-AP-150UL
10089-976EA 402 USD
10089-976
Anti-MARCH8 Rabbit Polyclonal Antibody
Antibodies
MARCH8(Membrane-associated RING finger protein 8) is also named as MIR, RNF178 and belongs to the RING zinc finger protein family. It functions as an E3 ubiquitin ligase for immune recognition-related molecules (e.g. major histocompatibility complex class I, B7-2, and ICAM-1). It plays a key role in controlling MHC class II surface expression by regulated ubiquitination of a lysine residue in the beta-chain.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IP: Mouse lung Tissue, 1:200-1:1000

Type: Primary
Antigen: MARCH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR