Anti-UBE2S Rabbit Polyclonal Antibody

Supplier: Proteintech
14115-1-AP-150UL
10081-258EA 402 USD
10081-258
Anti-UBE2S Rabbit Polyclonal Antibody
Antibodies
UBE2S(Ubiquitin-conjugating enzyme E2 S) is also named as E2EPF and belongs to the ubiquitin-conjugating enzyme family. It acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C) and a cell cycle-regulated ubiquitin ligase that controls progression through mitosis. Overexpression of UBE2S can increase tumor cell proliferation, invasion, and metastasis through the VHL-HIF pathway .

Western Blot: MDA-MB-453s Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: UBE2S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR