Anti-CNOT7 Rabbit Polyclonal Antibody

Supplier: Proteintech
14102-1-AP-150UL
10084-942EA 402 USD
10084-942
Anti-CNOT7 Rabbit Polyclonal Antibody
Antibodies
Cnot7, Ccr4-Not complex 7, is a component of CCR4-NOT complex and known to be a transcription factor and a modulator of mRNA degradation in yeast. Mammalian Cnot7 interacts with Tob and its family members, which is known as an inhibitor of BMP signaling through interfering with the Smad system and is involved in bone metabolism . It is one of at least 8 subunits that make up the CCR4-Not transcription complex, which is involved in negative regulation of the pol II directed transcription in yeast24 and may be involved in global gene regulation by modulation of TATAA-binding protein (TBP) activity .

Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: CNOT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR