Anti-SLC5A11 Rabbit Polyclonal Antibody

Supplier: Proteintech
14089-1-AP-150UL
10094-794EA 459.7 USD
10094-794
Anti-SLC5A11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC5A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR