Anti-SREBF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14088-1-AP-150UL
10095-154EA 614.88 USD
10095-154
Anti-SREBF1 Rabbit Polyclonal Antibody
Antibodies
SREBF1, also named as BHLHD1 and SREBP1, contains one basic helix-loop-helix (bHLH) domain and belongs to the SREBP family. It is a transcriptional activator required for lipid homeostasis. The SREBPs are synthesized as precursors anchored to endoplasmic reticulum (ER) membranes and complexed with SCAP. When the cellular cholesterol level is low, SREBP-SCAP complexes move to the Golgi apparatus, where SREBPs undergo a two-step proteolytic processing, leading to the release of the mature form, an N-terminal fragment, i.e, basic helix-loop-helix leucine zipper transcription factor. These factors enter the nucleus where they bind to sterol regulatory elements (SRE) in the promoter regions of a number of genes whose products mediate the synthesis of cholesterol and fatty acids. . This antibody can recognize the 125kd precursor form and the 68kd mature form of human SREBF1.

Western Blot: HeLa Cells, 1:200-1:2000; IP:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: SREBF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR