Anti-NINJ2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14085-1-AP-150UL
10091-106EA 402 USD
10091-106
Anti-NINJ2 Rabbit Polyclonal Antibody
Antibodies
NINJ2 (Ninjurin2) is a member of the ninjurin family of adhesion molecules which mediate cell-to-cell and cell-to-extracellular matrix interactions during development, differentiation, and regeneration of the peripheral nervous system. The gene encodes NINJ2 is located on chromosome 12p13. NINJ2 mRNA is widely expressed in adult human tissues, with highest level in bone marrow, followed by peripheral leukocytes, lung, and lymph nodes. In the peripheral nervous system, NINJ2 is expressed constitutively in mature sensory and enteric neurons. The expression of NINJ2 is upregulated after nerve injury in Schwann cells, suggesting that it may promote nerve regeneration. It may also play an important role in the pathogenesis of inflammatory disorder.

Western Blot:Transfected HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: NINJ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR