Anti-CNKSR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
14084-1-AP-150UL
10084-924EA 402 USD
10084-924
Anti-CNKSR3 Rabbit Polyclonal Antibody
Antibodies
Connector enhancer of kinase suppressor of ras 3 (CNKSR3) gene encodes CNK homolog protein 3 (CNK3). CNK proteins, originally identified in Drosophila, were described to regulate the RAS/ERK pathway at the level of the RAS protein kinase. CNKSR3, homologue of scaffold proteins involved in MAPK pathway regulation, is a direct target of mineralocorticoid receptor (MR). CNK3 coordinates the assembly of a multiprotein epithelial sodium channel (ENaC) -regulatory complex, and hence plays a central role in sodium homeostasis.

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: CNKSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR