Anti-SRPK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14073-1-AP-150UL
10095-182EA 402 USD
10095-182
Anti-SRPK1 Rabbit Polyclonal Antibody
Antibodies
SRPK1 (serine-arginine protein kinase 1) is a protein kinase that specifically phosphorylates proteins containing serine-arginine-rich domains. Its substrates include a family of SR proteins that are key regulators of mRNA AS (alternative splicing). SRPK1 may be localized both in the cytoplasm and nucleus and translocates between these compartments under various conditions (e.g. cell cycle and stress). It is able to phosphorylate SR proteins both in cytoplasm and nucleus (the significance of which remains incompletely understood) and thus regulate the activity of these proteins by affecting their ability to bind other proteins and/or RNA as well as their intracellular localization.. This protein has 2 isoforms produced by alternative splicing.

Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A; IF:SH-SY5Y Cells, 1:10-1:100

Type: Primary
Antigen: SRPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR