Anti-ACAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
14029-1-AP-150UL
10082-404EA 402 USD
10082-404
Anti-ACAP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:200-1:2000

Type: Primary
Antigen: ACAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR