Anti-MYO1D Rabbit Polyclonal Antibody

Supplier: Proteintech
14028-1-AP-150UL
10090-748EA 402 USD
10090-748
Anti-MYO1D Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: MYO1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR