Anti-PSMA8 Rabbit Polyclonal Antibody

Supplier: Proteintech
14022-1-AP-150UL
10092-900EA 402 USD
10092-900
Anti-PSMA8 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Testis Tissue, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: PSMA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR