Anti-AADACL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14021-1-AP-150UL
10082-276EA 402 USD
10082-276
Anti-AADACL1 Rabbit Polyclonal Antibody
Antibodies
AADACL1(Arylacetamide deacetylase-like 1) is also named as NCEH1, KIAA1363 and belongs to the 'GDXG' lipolytic enzyme family. The transmembrane enzyme, AADACL1, controls the production of the monoalkylglycerol ether (MAGE) class of NELs in cancer cells and acts as a 2-acetyl MAGE hydrolase and is likely the principal source for this activity in tumor cells. The full length protein has three glycosylation sites and can be N-glycosylated. It has 3 isoforms produced by alternative splicing.

Western Blot:COLO 320 Cells, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: AADACL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR