Anti-BACH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
14018-1-AP-150UL
10083-478EA 402 USD
10083-478
Anti-BACH1 Rabbit Polyclonal Antibody
Antibodies
BTB and CNC homology 1, basic leucine zipper transcription factor 1(BACH1) is a heme-regulated transcriptional regulator that acts as repressor or activator and a member of the AP-a superfamily. MafK belongs to the small Maf family that can funciton as transcriptional activators or repressors depending on the proteins with which they form heterdimers. BACH1 has crucial roles in both stress responses and transformation by H-RasV12

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: BACH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR