Anti-Cryptochrome 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13997-1-AP-150UL
10085-228EA 524.48 USD
10085-228
Anti-Cryptochrome 2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000

Type: Primary
Antigen: Cryptochrome 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR